ENA245487143 Enamine

2-(2-fluorophenyl)-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD07376769
  • Cas Number : 500605-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55137

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55137
Availability details for
Quantity
  • Availability
  • Add to Cart