ENA245487149 Enamine

[(2-ethyl-1,3-thiazol-4-yl)methyl](methyl)amine

  • MDL number MFCD09693919
  • Cas Number : 165316-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55143

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55143
Availability details for
Quantity
  • Availability
  • Add to Cart