ENA245487156 Enamine

1-{3-[2-(3-acetylphenoxy)ethoxy]phenyl}ethan-1-one

  • MDL number MFCD13196178
  • Cas Number : 112092-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117...119
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55152

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55152
Availability details for
Quantity
  • Availability
  • Add to Cart