ENA245487158 Enamine

sodium 2-[4-(4-fluorophenyl)-1,3-thiazol-2-yl]acetate

  • MDL number MFCD13196179
  • Cas Number : 1211774-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55154
Availability details for
Quantity
  • Availability
  • Add to Cart