ENA245487161 Enamine

N-(2-chloro-4-methylphenyl)-2-hydroxyacetamide

  • MDL number MFCD08443506
  • Cas Number : 926272-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55159

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55159
Availability details for
Quantity
  • Availability
  • Add to Cart