ENA245487167 Enamine

butanimidamide hydrochloride

  • MDL number MFCD00054341
  • Cas Number : 3020-81-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-108
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55168

Safety Information

signalword Warning
pcodes P264,P280,P305+P351+P338,P337+P313
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-55168
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart