ENA245487175 Enamine

1-(2-aminoethoxy)-2-methoxyethane

  • MDL number MFCD09032941
  • Cas Number : 31576-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55177

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55177
Availability details for
Quantity
  • Availability
  • Add to Cart