ENA245487176 Enamine

N-methyl-3-(propan-2-yl)aniline

  • MDL number MFCD09744932
  • Cas Number : 117802-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55178

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55178
Availability details for
Quantity
  • Availability
  • Add to Cart