ENA245487187 Enamine

3-methanesulfonyl-5-nitrobenzoic acid

  • MDL number MFCD12173027
  • Cas Number : 58123-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55198

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55198
Availability details for
Quantity
  • Availability
  • Add to Cart