ENA245487191 Enamine

2-(5-bromothiophen-2-yl)ethan-1-amine

  • MDL number MFCD04114538
  • Cas Number : 86423-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55202

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55202
Availability details for
Quantity
  • Availability
  • Add to Cart