ENA245487193 Enamine

2,2,2-trifluoroethyl N-(4-methyl-2-oxo-2H-chromen-7-yl)carbamate

  • MDL number MFCD13196191
  • Cas Number : 1210752-66-1
  • Molecular Weight : 301.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-55204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55204
Availability details for
Quantity
  • Availability
  • Add to Cart