ENA245487196 Enamine

1-(3-ethyl-1-benzofuran-2-yl)ethan-1-ol

  • MDL number MFCD08443412
  • Cas Number : 926272-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66...68
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55223

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55223
Availability details for
Quantity
  • Availability
  • Add to Cart