ENA245487449 Enamine

1-amino-3-(4-nitrophenoxy)propan-2-ol hydrochloride

  • MDL number MFCD13196204
  • Cas Number : 59117-67-8
  • Molecular Weight : 248.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-amino-3-(4-nitrophenoxy)-2-propanol hydrochloride
Inchi Code 1S/C9H12N2O4.ClH/c10-5-8(12)6-15-9-3-1-7(2-4-9)11(13)14;/h1-4,8,12H,5-6,10H2;1H
Inchi Key NETJZMHBDMYMOS-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-55286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55286
Availability details for
Quantity
  • Availability
  • Add to Cart