ENA245487453 Enamine

2-[4-(2-aminoethyl)phenoxy]ethan-1-ol hydrochloride

  • MDL number MFCD13196210
  • Cas Number : 1208889-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184...186
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55317

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55317
Availability details for
Quantity
  • Availability
  • Add to Cart