ENA245487457 Enamine

N-(2-methylpropyl)cyclohexanamine hydrochloride

  • MDL number MFCD13196214
  • Cas Number : 99863-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 275...277
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55353

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55353
Availability details for
Quantity
  • Availability
  • Add to Cart