ENA245487458 Enamine

2,2,2-trifluoroethyl N-(5-cyclohexyl-1,3,4-thiadiazol-2-yl)carbamate

  • MDL number MFCD13196215
  • Cas Number : 1209253-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55354
Availability details for
Quantity
  • Availability
  • Add to Cart