ENA245487459 Enamine

2-amino-N-(4-ethoxyphenyl)acetamide hydrochloride

  • MDL number MFCD13196216
  • Cas Number : 539-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55367

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55367
Availability details for
Quantity
  • Availability
  • Add to Cart