ENA245487463 Enamine

3-methoxy-4-sulfanylbenzaldehyde

  • MDL number MFCD13196219
  • Cas Number : 96495-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43...45
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55392

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55392
Availability details for
Quantity
  • Availability
  • Add to Cart