ENA245487464 Enamine

{[5-(3-fluorophenyl)thiophen-2-yl]methyl}(propan-2-yl)amine hydrochloride

  • MDL number MFCD13196220
  • Cas Number : 1208894-86-3
  • Molecular Weight : 285.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55394

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55394
Availability details for
Quantity
  • Availability
  • Add to Cart