ENA245487465 Enamine

{[5-(4-methylphenyl)thiophen-2-yl]methyl}(propan-2-yl)amine hydrochloride

  • MDL number MFCD13196221
  • Cas Number : 1209619-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55395
Availability details for
Quantity
  • Availability
  • Add to Cart