ENA245487471 Enamine

3-bromo-2-(chloromethyl)thiophene

  • MDL number MFCD13196227
  • Cas Number : 7353-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-55413

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55413
Availability details for
Quantity
  • Availability
  • Add to Cart