ENA245487487 Enamine

4-(phenoxymethyl)benzene-1-carboximidamide hydrochloride

  • MDL number MFCD13196250
  • Cas Number : 57928-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55644
Availability details for
Quantity
  • Availability
  • Add to Cart