ENA245487488 Enamine

2-[(3,4-dimethylphenyl)sulfanyl]ethanimidamide hydrochloride

  • MDL number MFCD13196251
  • Cas Number : 1210206-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55649

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55649
Availability details for
Quantity
  • Availability
  • Add to Cart