ENA245487493 Enamine

(4-methylcyclohexyl)hydrazine dihydrochloride

  • MDL number MFCD13196265
  • Cas Number : 1209337-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55731
Availability details for
Quantity
  • Availability
  • Add to Cart