ENA245487494 Enamine

(4-ethylcyclohexyl)hydrazine dihydrochloride

  • MDL number MFCD13196266
  • Cas Number : 1208490-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55734
Availability details for
Quantity
  • Availability
  • Add to Cart