ENA245487495 Enamine

(4-chlorobutanoyl)urea

  • MDL number MFCD13196268
  • Cas Number : 1208680-83-4
  • Molecular Weight : 164.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55741

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55741
Availability details for
Quantity
  • Availability
  • Add to Cart