ENA245487505 Enamine

2-(1,1-dioxo-4H-1$l^{6},2,4-benzothiadiazin-4-yl)acetic acid

  • MDL number MFCD13196279
  • Cas Number : 1209718-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55797
Availability details for
Quantity
  • Availability
  • Add to Cart