ENA245487507 Enamine

1-[(diethylcarbamoyl)methyl]-2-oxo-1,2-dihydropyridine-3-carboxylic acid

  • MDL number MFCD13196281
  • Cas Number : 1209796-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55810
Availability details for
Quantity
  • Availability
  • Add to Cart