ENA245487518 Enamine

5-fluoropyridine-3-sulfonyl chloride

  • MDL number MFCD13189070
  • Cas Number : 1060802-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35...37
Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55881

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-55881
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart