ENA245487519 Enamine

(2-methylcyclohexyl)hydrazine dihydrochloride

  • MDL number MFCD13196297
  • Cas Number : 1210687-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55901
Availability details for
Quantity
  • Availability
  • Add to Cart