ENA245487522 Enamine

N-methyl-N-(piperidin-4-yl)methanesulfonamide hydrochloride

  • MDL number MFCD13196300
  • Cas Number : 1148003-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-55913

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55913
Availability details for
Quantity
  • Availability
  • Add to Cart