ENA245487527 Enamine

3-(3-{[1,1'-biphenyl]-4-yl}-1-phenyl-1H-pyrazol-4-yl)propanoic acid

  • MDL number MFCD13196305
  • Cas Number : 1210822-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55930

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55930
Availability details for
Quantity
  • Availability
  • Add to Cart