ENA245487528 Enamine

1-(1,2,3,4-tetrahydronaphthalen-1-yl)-1H-pyrazol-5-amine dihydrochloride

  • MDL number MFCD13196306
  • Cas Number : 1211801-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55954

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55954
Availability details for
Quantity
  • Availability
  • Add to Cart