ENA245487530 Enamine

6-phenoxypyridine-3-carboximidamide dihydrochloride

  • MDL number MFCD13196309
  • Cas Number : 1208952-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55958

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55958
Availability details for
Quantity
  • Availability
  • Add to Cart