ENA245487531 Enamine

3-[(piperidin-1-yl)methyl]benzene-1-carboximidamide dihydrochloride

  • MDL number MFCD13196310
  • Cas Number : 1208849-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55960

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55960
Availability details for
Quantity
  • Availability
  • Add to Cart