ENA245487532 Enamine

ethyl 2-(1-benzyl-1H-imidazol-2-yl)-2-hydroxyacetate

  • MDL number MFCD13196311
  • Cas Number : 1210502-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55961
Availability details for
Quantity
  • Availability
  • Add to Cart