ENA245487533 Enamine

N-methyl-N-(piperidin-4-yl)acetamide hydrochloride

  • MDL number MFCD13196313
  • Cas Number : 550370-51-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208...210
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55963

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55963
Availability details for
Quantity
  • Availability
  • Add to Cart