ENA245487535 Enamine

3-(7-methoxynaphthalen-2-yl)-1-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD13196315
  • Cas Number : 1208697-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55983

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55983
Availability details for
Quantity
  • Availability
  • Add to Cart