ENA245487537 Enamine

1-(2-cyanoethyl)-3-(4-nitrophenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD13196318
  • Cas Number : 1211370-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55994

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55994
Availability details for
Quantity
  • Availability
  • Add to Cart