ENA245487539 Enamine

1-(2,4-dimethyl-1,3-thiazol-5-yl)ethan-1-ol

  • MDL number MFCD13196320
  • Cas Number : 89775-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56008
Availability details for
Quantity
  • Availability
  • Add to Cart