ENA245487542 Enamine

2-(dimethylamino)ethanimidamide dihydrochloride

  • MDL number MFCD13196324
  • Cas Number : 1210786-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-56067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56067
Availability details for
Quantity
  • Availability
  • Add to Cart