ENA245487680 Enamine

4-methoxy-3-{[2-(pyrrolidin-1-yl)ethyl]sulfamoyl}benzoic acid hydrochloride

  • MDL number MFCD13196257
  • Cas Number : 1203154-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229...231
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55697

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55697
Availability details for
Quantity
  • Availability
  • Add to Cart