ENA245487689 Enamine

N-[(3,4-dimethoxyphenyl)methyl]-1-methyl-4-nitro-1H-imidazol-5-amine

  • MDL number MFCD13196261
  • Cas Number : 1155610-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55706
Availability details for
Quantity
  • Availability
  • Add to Cart