ENA245487694 Enamine

(2-butoxypyridin-3-yl)methanamine

  • MDL number MFCD11625319
  • Cas Number : 851773-51-8
  • Molecular Weight : 180.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-55711

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55711
Availability details for
Quantity
  • Availability
  • Add to Cart