ENA245487698 Enamine

ethyl 3-[(pyridin-3-ylmethyl)amino]propanoate

  • MDL number MFCD10687258
  • Cas Number : 1040688-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55715

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55715
Availability details for
Quantity
  • Availability
  • Add to Cart