ENA245487708 Enamine

3,4,5-trifluorobenzonitrile

  • MDL number MFCD00074948
  • Cas Number : 134227-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-47
Purity 95
Flash Point 63
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55728

Safety Information

signalword Danger
pcodes P210,P240,P241,P280,P370+P378
MSDS https://www.enaminestore.com/catalog/EN300-55728
pictograms GHS02
hstatements H228
Availability details for
Quantity
  • Availability
  • Add to Cart