ENA245487714 Enamine

3-(2-methyl-1H-1,3-benzodiazol-1-yl)propan-1-ol

  • MDL number MFCD12186738
  • Cas Number : 577994-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55329

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55329
Availability details for
Quantity
  • Availability
  • Add to Cart