ENA245487720 Enamine

2-hydroxyethanimidamide hydrochloride

  • MDL number MFCD02856524
  • Cas Number : 54198-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-149
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55745
Availability details for
Quantity
  • Availability
  • Add to Cart