ENA245487721 Enamine

3-methyl-2-[(pyridin-4-yl)formamido]butanoic acid

  • MDL number MFCD00448262
  • Cas Number : 147218-37-9
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55747
Availability details for
Quantity
  • Availability
  • Add to Cart