ENA245487726 Enamine

4-(chloromethyl)-2-(2-chlorophenyl)-1,3-oxazole

  • MDL number MFCD11200991
  • Cas Number : 678165-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55753
Availability details for
Quantity
  • Availability
  • Add to Cart